Assay Details
Target Gene Details
Entrez Gene ID: | 23880 |
Gene Name: | FYN binding protein |
Gene Aliases: |
ADAP, B630013F22Rik, FYB-120/130, Slap130, p120/130, p120/p130 |
Location: |
Chr.15:6522797-6665608 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fyb | NM_001278269.1 | NP_001265198.1 | ||
| NM_011815.5 | NP_035945.1 | |||
| AF001863.1 | AAB62227.1 | |||
| AF061744.1 | AAD03267.1 | |||
| AK037454.1 | BAC29811.1 | |||
| AK038225.1 | ||||
| AK137907.1 | ||||
| AK172013.1 | BAE42776.1 | |||
| BC141352.1 |
More Information
Set Membership: |
|

Genomic Map