Assay Details
Target Gene Details
Entrez Gene ID: | 16529 |
Gene Name: | potassium channel, subfamily K, member 5 |
Gene Aliases: |
KCNK5b, TASK-2 |
Location: |
Chr.14:20140057-20181782 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Kcnk5 | NM_021542.4 | 5 | 1763 | NP_067517.1 |
| AF259395.1 | 5 | 1433 | AAF68668.1 | |
| AF319542.1 | 5 | 1502 | AAG35065.1 | |
| AK085330.1 | 5 | 1764 | BAC39423.1 | |
| AK132172.1 | 5 | 1791 | BAE21010.1 | |
| AK172608.1 | 5 | 1764 | BAE43089.1 | |
| BC034012.1 | 5 | 1765 | ||
| BC058164.1 | 5 | 1754 | AAH58164.1 |
More Information
Set Membership: |
|

Genomic Map