Assay Details
Target Gene Details
Entrez Gene ID: | 71145 |
Gene Name: | scavenger receptor class A, member 5 (putative) |
Gene Aliases: |
4932433F15Rik, 4933425F03Rik, AV278087, Tesr |
Location: |
Chr.14:65666403-65764826 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Scara5 | NM_028903.2 | 9 | 2735 | NP_083179.2 |
| AK133176.1 | 9 | 2736 | BAE21542.1 | |
| BC016096.1 | 9 | 2711 | AAH16096.1 | |
| BC023907.1 | 9 | 2651 | AAH23907.1 | |
| BC026758.1 | 6 | 1761 | AAH26758.1 | |
| BC032159.1 | 9 | 2691 | AAH32159.1 |

Genomic Map