Assay Details
Target Gene Details
Entrez Gene ID: | 73569 |
Gene Name: | vestigial like 3 (Drosophila) |
Gene Aliases: |
1700110N18Rik, 4832416J22, C80713, Vgl-3, Vito-2 |
Location: |
Chr.16:65815017-65866368 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Vgll3 | NM_028572.1 | 2 | 305 | NP_082848.1 |
More Information
Set Membership: |
|

Genomic Map