Assay Details
Target Gene Details
Entrez Gene ID: | 269037 |
Gene Name: | CBP80/20-dependent translation initiation factor |
Gene Aliases: |
Gm672 |
Location: |
Chr.18:75431205-75697696 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ctif | NM_201354.2 | 8 | 1158 | NP_958742.2 |
| BC058104.1 | 8 | 1016 | AAH58104.1 |
Target Gene Details
Entrez Gene ID: | 100038353 |
Gene Name: | predicted gene 10532 |
Gene Aliases: |
ENSMUSG00000073534 |
Location: |
Chr.18:75514645-75522806 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gm10532 | NR_045879.1 | 4 | 1122 | |
| AK133424.1 | 4 | 1123 | BAE21650.1 |
More Information
Set Membership: |
|

Genomic Map