Assay Details
Target Gene Details
Entrez Gene ID: | 268977 |
Gene Name: | latent transforming growth factor beta binding protein 1 |
Gene Aliases: |
9430031G15Rik, 9830146M04, Ltbp-1, Tgfb, b2b1000Clo |
Location: |
Chr.17:75005529-75392967 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ltbp1 | NM_019919.3 | 1 | 278 | NP_064303.2 |
| AF022889.1 | 1 | 243 | AAC33307.1 | |
| AK053964.1 | BAC35603.1 | |||
| BB613838.1 | 1 | 279 |
More Information
Set Membership: |
|

Genomic Map