Assay Details
Target Gene Details
Entrez Gene ID: | 67180 |
Gene Name: | Yip1 domain family, member 5 |
Gene Aliases: |
2610311I19Rik, AA408236, Yip1a |
Location: |
Chr.18:40204865-40219399 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Yipf5 | NM_023311.3 | 6 | 2068 | NP_075800.1 |
| AK153123.1 | 6 | 2027 | BAE31738.1 | |
| AV120181.2 | 1 | 752 |

Genomic Map