Assay Details
Target Gene Details
Entrez Gene ID: | 14156 |
Gene Name: | flap structure specific endonuclease 1 |
Gene Aliases: |
AW538437, FEN-1 |
Location: |
Chr.19:10199132-10204187 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fen1 | NM_001271614.1 | 1 | 89 | NP_001258543.1 |
| NM_001271615.1 | 1 | 89 | NP_001258544.1 | |
| NM_007999.4 | 1 | 89 | NP_032025.2 | |
| AK042942.1 | BAC31412.1 | |||
| AK046256.1 | 1 | 85 | BAC32660.1 | |
| AK076977.1 | 1 | 63 | BAC36544.1 | |
| AK078778.1 | 1 | 76 | BAC37390.1 | |
| AK153889.1 | 1 | 56 | BAE32236.1 | |
| AK158431.1 | 1 | 62 | ||
| AK161047.1 | 1 | 69 | BAE36169.1 | |
| AK166948.1 | 1 | 57 | BAE39135.1 | |
| AK168731.1 | 1 | 81 | BAE40572.1 | |
| BC010203.1 | AAH10203.1 | |||
| BC027295.1 | 1 | 69 | AAH27295.1 | |
| BY072885.1 | 1 | 63 | ||
| BY075857.1 | 1 | 90 | ||
| BY084492.1 | 1 | 90 | ||
| BY162898.1 | 1 | 82 |
More Information
Set Membership: |
|

Genomic Map