Assay Details
Target Gene Details
Entrez Gene ID: | 209268 |
Gene Name: | immunoglobulin superfamily, member 1 |
Gene Aliases: |
5330413N23, 5530402E03, AI747649, InhBP/p120, mKIAA0364 |
Location: |
Chr.X:49782536-49797906 on Build GRCm38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Igsf1 | NM_177591.4 | 12 | 2118 | NP_808259.2 |
| NM_183335.2 | 11 | 1982 | NP_899178.2 | |
| NM_183336.2 | 3 | 372 | NP_899179.1 | |
| AK030701.1 | 3 | 374 | ||
| AK076427.1 | 3 | 365 | BAC36338.1 | |
| AK129126.1 | 12 | 2100 | BAC97936.1 | |
| AY227771.1 | 12 | 2112 | AAP57079.1 | |
| AY227773.1 | 11 | 1982 | AAP57081.1 | |
| AY227774.1 | 3 | 367 | AAP57082.1 | |
| BC119264.1 | 3 | 335 | ||
| BC119266.1 | 3 | 335 |
More Information
Set Membership: |
|

Genomic Map