Assay Details
Target Gene Details
Entrez Gene ID: | 13098 |
Gene Name: | cytochrome P450, family 2, subfamily c, polypeptide 39 |
Gene Aliases: |
CYPIIC39 |
Location: |
Chr.19:39510822-39568529 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cyp2c39 | NM_010003.2 | 9 | 1505 | NP_034133.2 |
| AF047726.1 | 9 | 1456 | AAD13721.1 | |
| BC057911.1 | 9 | 1466 | AAH57911.1 | |
| BC139290.1 | 9 | 1505 | ||
| BC139291.1 | 9 | 1505 |
More Information
Set Membership: |
|

Genomic Map