Assay Details
Target Gene Details
Entrez Gene ID: | 320707 |
Gene Name: | ATPase, Ca++ transporting, plasma membrane 3 |
Gene Aliases: |
6430519O13Rik, Pmca3 |
Location: |
Chr.X:73503023-73573275 on Build GRCm38 |
Assay Gene Location: | Within Exon 21 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Atp2b3 | NM_177236.4 | 21 | 3838 | NP_796210.2 |
| AK045516.1 | 21 | 3837 | ||
| BC118975.1 | 20 | 3570 | ||
| BC118976.1 | 20 | 3532 |
More Information
Set Membership: |
|

Genomic Map