Assay Details
Target Gene Details
Entrez Gene ID: | 240216 |
Gene Name: | Riken cDNA E230025N22 gene |
Gene Aliases: |
EG240216 |
Location: |
Chr.18:36683078-36696091 on Build GRCm38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| E230025N22Rik |
Target Gene Details
Entrez Gene ID: | 67843 |
Gene Name: | solute carrier family 35, member A4 |
Gene Aliases: |
2610030J16Rik |
Location: |
Chr.18:36679215-36683862 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc35a4 | NM_001083317.1 | 4 | 2495 | NP_001076786.1 |
| NM_026404.2 | 3 | 2603 | NP_080680.2 | |
| AK011633.1 | 3 | 2227 | BAB27747.1 | |
| AK031132.1 | 3 | 2227 | BAC27268.1 | |
| AK145812.1 | 3 | 2227 | BAE26666.1 | |
| AK164797.1 | 4 | 2119 | BAE37923.1 | |
| AK170045.1 | 4 | 2132 | BAE41529.1 | |
| BC006050.1 | 2 | 2204 | AAH06050.1 | |
| BP765876.1 | 1 | 259 |
More Information
Set Membership: |
|

Genomic Map