Assay Details
Target Gene Details
Entrez Gene ID: | 67738 |
Gene Name: | peptidylprolyl isomerase D (cyclophilin D) |
Gene Aliases: |
4930564J03Rik, CYP-40, Ppidl, Ppif |
Location: |
Chr.3:79591389-79603650 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 7 - Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ppid | NM_026352.3 | NP_080628.1 | ||
| AK003402.1 | BAB22767.1 | |||
| AK012028.1 | BAC25351.1 | |||
| AK013919.1 | BAB29056.1 | |||
| AK016221.1 | ||||
| AK051597.1 | BAC34686.1 | |||
| AK142413.1 | ||||
| AK150520.1 | BAE29632.1 | |||
| AK151091.1 | BAE30104.1 | |||
| BC011499.1 | AAH11499.1 | |||
| BC019778.1 | AAH19778.1 |
More Information
Set Membership: |
|

Genomic Map