Assay Details
Target Gene Details
Entrez Gene ID: | 67578 |
Gene Name: | protein associated with topoisomerase II homolog 2 (yeast) |
Gene Aliases: |
4930424G05Rik, Pat1a |
Location: |
Chr.2:122120103-122186189 on Build GRCm38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Patl2 | NM_026251.2 | NP_080527.2 | ||
| AK015195.1 | BAB29742.1 | |||
| BY714747.1 |
Target Gene Details
Entrez Gene ID: | 70928 |
Gene Name: | tripartite motif-containing 69 |
Gene Aliases: |
4921519C19Rik, Rnf36, Trif, Trimless |
Location: |
Chr.2:122160700-122179027 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Trim69 | NM_080510.2 | NP_536771.2 | ||
| AF334958.1 | AAL41031.1 | |||
| AK014923.1 | ||||
| BC050815.1 | AAH50815.1 |
More Information
Set Membership: |
|

Genomic Map