Assay Details
Target Gene Details
Entrez Gene ID: | 72873 |
Gene Name: | coiled-coil domain containing 176 |
Gene Aliases: |
2900006K08Rik, AI427784 |
Location: |
Chr.12:84409068-84441763 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ccdc176 | NM_028377.3 | NP_082653.2 | ||
| AK133381.1 | BAE21625.1 | |||
| BC151099.1 | ||||
| BC151103.1 |

Genomic Map