Assay Details
Target Gene Details
Entrez Gene ID: | 66884 |
Gene Name: | amyloid beta precursor protein (cytoplasmic tail) binding protein 2 |
Gene Aliases: |
1300003O07Rik, AI465480, PAT1 |
Location: |
Chr.11:85191308-85235120 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Appbp2 | NM_025825.3 | 2 | 486 | NP_080101.1 |
| AK004888.1 | 2 | 497 | ||
| AK005440.1 | 2 | 192 | BAB24034.1 | |
| AK133853.1 | 2 | 478 | ||
| AK153962.1 | 1 | 3574 | ||
| AK177656.1 | 1 | 50 | ||
| AK182121.1 | 1 | 50 | ||
| BC018442.1 | 2 | 464 | AAH18442.1 | |
| BC019152.1 | 2 | 487 | AAH19152.1 |

Genomic Map