Assay Details
Target Gene Details
Entrez Gene ID: | 13136 |
Gene Name: | CD55 antigen |
Gene Aliases: |
Daf, Daf-GPI, Daf1, GPI-DAF |
Location: |
Chr.1:130437196-130463012 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cd55 | NM_010016.2 | NP_034146.2 | ||
| AK030285.1 | ||||
| AK160994.1 | BAE36139.1 | |||
| BC011314.1 | AAH11314.1 | |||
| BU515742.1 | ||||
| CT010328.1 | CAJ18536.1 | |||
| D63679.1 | BAA09830.1 | |||
| L41366.1 | AAB00091.1 |

Genomic Map