Assay Details
Target Gene Details
Entrez Gene ID: | 12268 |
Gene Name: | complement component 4B (Chido blood group) |
Gene Aliases: |
C4, Ss |
Location: |
Chr.17:34728046-34743901 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 11 - Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| C4b | NM_009780.2 | NP_033910.2 | ||
| AK157954.1 | BAE34280.1 | |||
| AK158256.1 | BAE34429.1 | |||
| AK158475.1 | BAE34531.1 | |||
| AK171644.1 | BAE42584.1 | |||
| AK172637.1 | ||||
| BC067394.1 | AAH67394.1 | |||
| BC067409.1 | AAH67409.1 | |||
| BC150743.1 | ||||
| BC158005.1 | ||||
| M11729.1 | AAA39506.1 |
Target Gene Details
Entrez Gene ID: | 81877 |
Gene Name: | tenascin XB |
Gene Aliases: |
Tn-mhc, Tnx |
Location: |
Chr.17:34659788-34795460 on Build GRCm38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tnxb |
More Information
Set Membership: |
|

Genomic Map