Assay Details
Target Gene Details
Entrez Gene ID: | 20688 |
Gene Name: | trans-acting transcription factor 4 |
Gene Aliases: |
5730497N03Rik, HF-1b, HF1-b |
Location: |
Chr.12:118231686-118301440 on Build GRCm38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sp4 | NM_001166385.2 | NP_001159857.2 | ||
| NM_009239.4 | NP_033265.4 | |||
| AB221633.1 | ||||
| BC076630.1 | AAH76630.1 | |||
| U62522.1 | AAC52653.1 |
More Information
Set Membership: |
|

Genomic Map