Assay Details
Target Gene Details
Entrez Gene ID: | 26987 |
Gene Name: | eukaryotic translation initiation factor 4E member 2 |
Gene Aliases: |
2700069E09Rik, AI036339, AV129531, D0H0S6743E, Eif4el3 |
Location: |
Chr.1:87213910-87240488 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Eif4e2 | NM_001039169.1 | NP_001034258.1 | ||
| NM_001039170.1 | NP_001034259.1 | |||
| NM_023314.3 | NP_075803.2 | |||
| AF068116.1 | AAC19373.1 | |||
| AK017221.1 | ||||
| AK018516.1 | BAB31251.1 | |||
| AK032965.1 | BAC28102.1 | |||
| AK138055.1 | BAE23544.1 | |||
| AK150495.1 | BAE29610.1 | |||
| AK199854.1 | ||||
| BC028903.1 | ||||
| BC045153.1 | AAH45153.1 | |||
| BC049077.1 | AAH49077.1 | |||
| U01137.1 |
More Information
Set Membership: |
|

Genomic Map