Assay Details
Target Gene Details
Entrez Gene ID: | 75597 |
Gene Name: | NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, assembly factor 2 |
Gene Aliases: |
1810058I14Rik, C86051, Ndufa12l, mimitin |
Location: |
Chr.13:108052589-108158654 on Build GRCm38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ndufaf2 | NM_001127346.1 | NP_001120818.1 | ||
| AB183434.1 | BAD91206.1 | |||
| AK007894.1 | ||||
| AK166492.1 | BAE38806.1 | |||
| AY262012.1 | AAP33449.1 | |||
| BC116975.1 | ||||
| BC116977.1 | ||||
| BY251841.1 |
More Information
Set Membership: |
|

Genomic Map