Assay Details
Target Gene Details
Entrez Gene ID: | 26384 |
Gene Name: | glucosamine-6-phosphate deaminase 1 |
Gene Aliases: |
GNPDA, Gnp1, Gnpi, oscillin |
Location: |
Chr.18:38327537-38338993 on Build GRCm38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gnpda1 | NM_011937.2 | NP_036067.2 | ||
| AF088903.1 | AAC36739.1 | |||
| AF160355.1 | AAD42233.1 | |||
| AK080610.1 | ||||
| AK166069.1 | ||||
| AK172879.1 | BAD32157.1 | |||
| BC014800.1 | AAH14800.1 | |||
| BY092839.1 | ||||
| CJ048539.1 |
More Information
Set Membership: |
|

Genomic Map