Assay Details
Target Gene Details
Entrez Gene ID: | 16558 |
Gene Name: | kinesin family member 16B |
Gene Aliases: |
8430434E15Rik, AA623607, AI789011, C80253, C80902, mKIAA1590 |
Location: |
Chr.2:142618334-142901595 on Build GRCm38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Kif16b | NM_001081133.2 | NP_001074602.1 | ||
| AB001423.1 | BAA22383.1 | |||
| AB610968.1 | ||||
| CN664558.1 |
More Information
Set Membership: |
|

Genomic Map