Assay Details
Target Gene Details
Entrez Gene ID: | 16911 |
Gene Name: | LIM domain only 4 |
Gene Aliases: |
A730077C12Rik, Crp3, Etohi4 |
Location: |
Chr.3:144188530-144205255 on Build GRCm38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Lmo4 | NM_001161769.1 | NP_001155241.1 | ||
| NM_001161770.1 | NP_001155242.1 | |||
| NM_010723.3 | NP_034853.1 | |||
| AF096996.1 | AAC83789.1 | |||
| AF102817.1 | AAC98510.1 | |||
| AK080534.1 | BAC37938.1 | |||
| AK138278.1 | BAE23609.1 | |||
| AK159590.1 | BAE35210.1 | |||
| BC003488.1 | AAH03488.1 | |||
| BC010278.1 | AAH10278.3 | |||
| BC100382.1 | AAI00383.1 |
More Information
Set Membership: |
|

Genomic Map