Assay Details
| Gene Symbol: FER1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_120238.4 | NP_195780.1 | 5 - 6 | 1065 | 121 | |
| GenBank mRNA | AF326869.1 | - | 5 - 6 | 677 | 121 | |
| AF339691.1 | - | 5 - 6 | 608 | 121 | ||
| AF412065.1 | - | 5 - 6 | 677 | 121 | ||
| AK226433.1 | - | 5 - 6 | 680 | 121 | ||
| X94248.1 | - | 646 | 121 | |||
More Information
Set Membership: |
|

FER1
Genomic Map