Assay Details
| Gene Symbol: FER1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_120238.4 | NP_195780.1 | 7 - 8 | 1188 | 84 | |
| GenBank mRNA | AF326869.1 | - | 7 - 8 | 800 | 84 | |
| AF339691.1 | - | 7 - 8 | 731 | 84 | ||
| AF412065.1 | - | 7 - 8 | 800 | 84 | ||
| AK226433.1 | - | 7 - 8 | 803 | 84 | ||
| X94248.1 | - | 769 | 84 | |||
More Information
Set Membership: |
|

FER1
Genomic Map