Assay Details
| Gene Symbol: ABCB29 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_120473.3 | NP_196011.1 | 8 - 9 | 1698 | 101 | |
| GenBank mRNA | AY059727.1 | - | 8 - 9 | 1693 | 101 | |
| AY091365.1 | - | 8 - 9 | 1631 | 101 | ||
More Information
Set Membership: |
|

ABCB29
Genomic Map