Assay Details
| Gene Symbol: XK-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_127708.3 | NP_179732.2 | 8 - 9 | 1319 | 82 | |
| NM_201776.3 | NP_973505.1 | 8 - 9 | 1306 | 82 | ||
| GenBank mRNA | AK175657.1 | - | 8 - 9 | 1240 | 82 | |
| AK176783.1 | - | 8 - 9 | 1240 | 82 | ||
| AY136397.1 | - | 8 - 9 | 1242 | 82 | ||
| BT000210.1 | - | 8 - 9 | 1227 | 82 | ||
More Information
Set Membership: |
|

XK-1
Genomic Map