Assay Details
| Gene Symbol: PHS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036856.2 | NP_001031933.1 | 1 - 2 | 245 | 82 | |
| NM_122277.2 | NP_197761.2 | 2 - 3 | 143 | 82 | ||
| NM_180735.4 | NP_851066.2 | 2 - 3 | 227 | 82 | ||
| GenBank mRNA | AB161693.1 | - | 2 - 3 | 194 | 82 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_122277.2 transcripts.
Set Membership: |
|

PHS1
Genomic Map