Assay Details
| Gene Symbol: HDA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_122505.4 | NP_568480.2 | 1 - 2 | 359 | 109 | |
| NM_180747.2 | NP_851078.1 | 1 - 2 | 313 | 109 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

HDA2| Gene Symbol: HDA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_122505.4 | NP_568480.2 | 1 - 2 | 359 | 109 | |
| NM_180747.2 | NP_851078.1 | 1 - 2 | 313 | 109 | ||
Set Membership: |
|