Assay Details
| Gene Symbol: RGD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001203156.1 | NP_001190085.1 | 12 - 13 | 1966 | 85 | |
| NM_115288.8 | NP_190996.3 | 11 - 12 | 1997 | 85 | ||
| GenBank mRNA | AB440793.1 | - | 10 - 11 | 1735 | 85 | |
More Information
Set Membership: |
|

RGD3
Genomic Map