Assay Details
| Gene Symbol: GR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_115323.4 | NP_191026.1 | 7 - 8 | 1463 | 90 | |
| GenBank mRNA | BT020536.1 | - | 7 - 8 | 1369 | 90 | |
| BT030335.1 | - | 7 - 8 | 1352 | 90 | ||
| D14049.1 | - | 7 - 8 | 1370 | 90 | ||
More Information
Set Membership: |
|

GR
Genomic Map