Assay Details
| Gene Symbol: BCCP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_121557.4 | NP_568316.1 | 1 - 2 | 518 | 65 | |
| GenBank mRNA | AF223948.1 | - | 1 - 2 | 219 | 65 | |
| AY086528.1 | - | 1 - 2 | 309 | 65 | ||
| BT028902.1 | - | 1 - 2 | 126 | 65 | ||
More Information
Set Membership: |
|

BCCP2
Genomic Map