Assay Details
| Gene Symbol: TSA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_104119.3 | NP_564607.1 | 5 - 6 | 368 | 84 | |
| NM_179466.3 | NP_849797.1 | 5 - 6 | 493 | 84 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

TSA1| Gene Symbol: TSA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_104119.3 | NP_564607.1 | 5 - 6 | 368 | 84 | |
| NM_179466.3 | NP_849797.1 | 5 - 6 | 493 | 84 | ||
Set Membership: |
|