Assay Details
| Gene Symbol: CLC-F | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_104438.4 | NP_564698.1 | 7 - 8 | 2489 | 69 | |
| NM_179482.3 | NP_849813.1 | 7 - 8 | 2489 | 69 | ||
| GenBank mRNA | AF366368.1 | - | 7 - 8 | 2351 | 69 | |
| AJ303348.1 | - | 7 - 8 | 2355 | 69 | ||
More Information
Set Membership: |
|

CLC-F
Genomic Map