Assay Details
| Gene Symbol: MGL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_105141.3 | NP_176647.1 | 2 - 3 | 921 | 107 | |
| GenBank mRNA | AF428413.1 | - | 2 - 3 | 830 | 107 | |
| AK226375.1 | - | 2 - 3 | 836 | 107 | ||
| AY054546.1 | - | 2 - 3 | 830 | 107 | ||
| BT006588.1 | - | 2 - 3 | 774 | 107 | ||
More Information
Set Membership: |
|

MGL
Genomic Map