Assay Details
| Gene Symbol: ARD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001340961.1 | NP_001328513.1 | 6 - 7 | 800 | 64 | |
| NM_117556.4 | NP_567443.1 | 6 - 7 | 636 | 64 | ||
| GenBank mRNA | AK118285.1 | - | 6 - 7 | 603 | 64 | |
| AK227847.1 | - | 6 - 7 | 604 | 64 | ||
| BT004801.1 | - | 6 - 7 | 540 | 64 | ||
| BT005440.1 | - | 6 - 7 | 540 | 64 | ||
More Information
Set Membership: |
|

ARD1
Genomic Map