Assay Details
| Gene Symbol: ABO6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_147837.4 | NP_680142.2 | 16 - 17 | 2940 | 66 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ABO6| Gene Symbol: ABO6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_147837.4 | NP_680142.2 | 16 - 17 | 2940 | 66 | |
Set Membership: |
|