Assay Details
| Gene Symbol: ADA2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001084653.2 | NP_001078122.2 | 4 - 5 | 495 | 84 | |
| NM_001202911.1 | NP_001189840.1 | 6 - 7 | 611 | 84 | ||
| NM_111653.2 | NP_566317.1 | 6 - 7 | 590 | 84 | ||
| NM_202522.1 | NP_974251.1 | 6 - 7 | 676 | 84 | ||
| GenBank mRNA | AF338769.1 | - | 6 - 7 | 566 | 84 | |
| AF360246.1 | - | 6 - 7 | 590 | 84 | ||
| AY040045.1 | - | 6 - 7 | 557 | 84 | ||
More Information
Set Membership: |
|

ADA2A
Genomic Map