Assay Details
| Gene Symbol: UBP13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001202935.1 | NP_001189864.1 | 15 - 16 | 2099 | 85 | |
| NM_001337961.1 | NP_001319526.1 | 15 - 16 | 2099 | 85 | ||
| NM_001337962.1 | NP_001326292.1 | 15 - 16 | 2102 | 85 | ||
| NM_112024.6 | NP_187797.3 | 15 - 16 | 2028 | 85 | ||
| GenBank mRNA | BT002760.1 | - | 15 - 16 | 1829 | 85 | |
More Information
Set Membership: |
|

UBP13
Genomic Map