Assay Details
| Gene Symbol: TYRDC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001084992.1 | NP_001078461.1 | 11 - 12 | 1518 | 81 | |
| NM_001203932.1 | NP_001190861.1 | 11 - 12 | 1646 | 81 | ||
| NM_001203933.2 | NP_001190862.1 | 12 - 13 | 1599 | 81 | ||
| NM_001341927.1 | NP_001329473.1 | 11 - 12 | 1695 | 81 | ||
| NM_001341928.1 | NP_001329474.1 | 9 - 10 | 2040 | 81 | ||
| NM_001341929.1 | NP_001329472.1 | 10 - 11 | 1937 | 81 | ||
| NM_119010.3 | NP_194597.1 | 11 - 12 | 1512 | 81 | ||
More Information
Set Membership: |
|

TYRDC
Genomic Map