Assay Details
| Gene Symbol: MGE1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_124902.3 | NP_200331.1 | 1 - 2 | 298 | 63 | |
| GenBank mRNA | AY042852.1 | - | 1 - 2 | 214 | 63 | |
| AY081551.1 | - | 1 - 2 | 143 | 63 | ||
More Information
Set Membership: |
|

MGE1
Genomic Map