Assay Details
| Gene Symbol: emb2742 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_112102.3 | NP_187873.1 | 6 - 7 | 584 | 75 | |
| GenBank mRNA | AY093110.1 | - | 6 - 7 | 495 | 75 | |
| BT008360.1 | - | 5 - 6 | 446 | 75 | ||
More Information
Set Membership: |
|

emb2742
Genomic Map