Assay Details
| Gene Symbol: HB-8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_119441.5 | NP_195014.1 | 12 - 13 | 1800 | 67 | |
| GenBank mRNA | AJ441292.1 | - | 11 - 12 | 1389 | 67 | |
| AY099631.1 | - | 12 - 13 | 1775 | 67 | ||
| BT008798.1 | - | 11 - 12 | 1389 | 67 | ||
More Information
Set Membership: |
|

HB-8
Genomic Map