Assay Details
| Gene Symbol: HDA14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_119501.4 | NP_567921.1 | 4 - 5 | 525 | 95 | |
| GenBank mRNA | AY052234.1 | - | 4 - 5 | 479 | 95 | |
| AY113069.1 | - | 4 - 5 | 414 | 95 | ||
More Information
Set Membership: |
|

HDA14
Genomic Map