Assay Details
| Gene Symbol: HPT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001335615.1 | NP_001325303.1 | 10 - 11 | 1301 | 88 | |
| NM_179653.4 | NP_849984.1 | 11 - 12 | 1328 | 88 | ||
| GenBank mRNA | AF324344.1 | - | 11 - 12 | 1150 | 88 | |
| AY063893.1 | - | 11 - 12 | 1180 | 88 | ||
| AY089963.1 | - | 11 - 12 | 1036 | 88 | ||
| AY113993.1 | - | 11 - 12 | 1036 | 88 | ||
| GU198365.1 | - | 11 - 12 | 1114 | 88 | ||
More Information
Set Membership: |
|

HPT1
Genomic Map