Assay Details
| Gene Symbol: AT3G16060 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_112476.3 | NP_566534.1 | 10 - 11 | 2042 | 91 | |
| GenBank mRNA | AY052352.1 | - | 10 - 11 | 1924 | 91 | |
| AY059654.1 | - | 10 - 11 | 1811 | 91 | ||
More Information
Set Membership: |
|

AT3G16060
Genomic Map