Assay Details
| Gene Symbol: DAC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_112674.4 | NP_566593.1 | 1 - 2 | 159 | 118 | |
| GenBank mRNA | AY039550.1 | - | 1 - 2 | 128 | 118 | |
| AY093751.1 | - | 1 - 2 | 95 | 118 | ||
More Information
Set Membership: |
|

DAC
Genomic Map