Assay Details
| Gene Symbol: ATSYTF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_112722.3 | NP_566607.1 | 2 - 3 | 1186 | 75 | |
| GenBank mRNA | AY056799.1 | - | 2 - 3 | 655 | 75 | |
| AY133527.1 | - | 2 - 3 | 268 | 75 | ||
More Information
Set Membership: |
|

ATSYTF
Genomic Map