Assay Details
| Gene Symbol: CYCB1;1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_119913.3 | NP_195465.1 | 3 - 4 | 506 | 117 | |
| GenBank mRNA | BT029006.1 | - | 3 - 4 | 406 | 117 | |
More Information
Set Membership: |
|

CYCB1;1
Genomic Map